VWR® Ultra-High Performance, 离心管,超高性能,光敏感
供应商: VWR Collection
VWRI525-1115EA 797 CNY
VWRI525-1115 VWRI525-1116 VWRI525-1117 VWRI525-1118 VWRI525-1119 VWRI525-1120
VWR® Ultra-High Performance, 离心管,超高性能,光敏感
管子 离心管
这些防漏离心管是由深色琥珀色PP材质制成的,可通过阻挡紫外线来保护光敏样品。
  • Easy-to-read black graduations in ±2% increments
  • Contains a large, white frosted writing area
  • RNase-, DNase-, and endotoxin-free
  • Autoclavable at 121 °C and freezable to –80 °C
  • Can be centrifuged up to 12500×g
  • Sterile tubes are sterilized by irradiation
  • Blocks 99% of light with wavelength less than 550 nm
  • During the freezing process, it is better to use plastic rack
  • During high-speed centrifugation, it is better to use suitable rotor adapters
这些无菌试管具有打印的标记区域,可快速方便地识别样品。 包括绿色高密度聚乙烯盖。 试管有15或50毫升大小,并提供多种包装形式。 所有的管和盖都可More Product Information