VWR® IS High Capacity Balances
供应商: VWR Collection
VWRI611-4530EA 16025 CNY
VWRI611-4530 VWRI611-4549 VWRI611-4550 VWRI611-4536 VWRI611-4528 VWRI611-4537 VWRI611-4529 VWRI611-4539 VWRI611-4557 VWRI611-4558 VWRI611-4559 VWRI611-4540 VWRI611-4538 VWRI611-4556 VWRI611-4547 VWRI611-4548 VWRI611-4520 VWRI611-4546 VWRI611-4946 VWRI611-4947 VWRI611-4948 VWRI611-4949 VWRI611-4950 VWRI611-4951
VWR® IS High Capacity Balances
天平
The IS range of high capacity balances are robust, reliable and ensure high performance. Models are equipped with either a backlit LCD with adjustable contrast, a large graphic display or a large 5" colour touch screen, the balances have been designed to deliver top performance at competitive prices.
  • Electromagnetic force restoration weighing system (for 0,01 g resolution)
  • High readability load cell weighing system (for 0,1 g resolution)
  • Solidly built with an ergonomic keypad
  • External calibration
  • Measuring units are selectable and customisable
  • Hook for below balance weighing can be used for weighing large samples or solid density determination (software included)
  • Multiple languages: English, German, French, Italian, Spanish and Portuguese (apart from models with LCD)
Models with a large dot matrix graphic display for easy reading and user friendly operation, have an extensive formulation (recipe) database, GLP/ISO record of last stored calibration data, and various advanced embedded features that make them perfect for laboratory use. Functions include compound formulation and quality control applications. Units feature an RS232 serial interface and are ideal for check weighing, percentage weighing, piece counting, totalising, dynamic weighing and max. load determination (peak hold).
Models with a large 5" colour touch screen (800×480 display) have the same features as the above graphic display models, and in addition give easy and intuitive access to all the functions thanks to the icon-driven menu. This gives user-friendly access to all the balance’s advanced applications and internal database, ensuring more efficiency, productivity and interactivity. These advanced models are suitable for multi-users as they have password protection functionality and feature a USB port for importing/exporting data from the database. Functions include advanced piece counting with statistics report and textile function to measure fibres with statistics report. Touch screen models include statistics capabilities for all the functions included with graphic display models, they also have database capabilities that other models do not have.
Delivery information: Supplied with protective plastic cover.