乙二胺四乙酸钠盐 0.01 M (M/100), StandARd® 滴定液, Macron Fine Chemicals™
供应商: Avantor Performance Materials
MACRH105-05EA 889 CNY
MACRH105-05 MACRH105-07
乙二胺四乙酸钠盐 0.01 M (M/100), StandARd® 滴定液, Macron Fine Chemicals™
乙二胺四乙酸钠盐
For calcium hardness determination.
分子式: C₁₀H₁₄N₂Na₂O₈
存储条件: Ambient
MDL编号: MFCD00070672
CAS号: 139-33-3
EINECS: 205-358-3
现在订购
产品技术规格
For Laboratory,Research,or Manufacturing Use
Molarity 0.0095 - 0.0105