乙二胺四乙酸钠盐 0.1 M (M/10), StandARd® volumetric solution, Macron Fine Chemicals™
供应商: Avantor Performance Materials
Synonyms: 四乙酸二氨基乙烯, EDTA
MACRH100-07EA 2013 CNY
MACRH100-07 MACRH100-05
乙二胺四乙酸钠盐 0.1 M (M/10), StandARd® volumetric solution, Macron Fine Chemicals™
乙二胺四乙酸钠盐
分子式: C₁₀H₁₄N₂Na₂O₈
分子量: 336.21 g/mol
存储条件: Ambient
MDL编号: MFCD00070672
CAS号: 139-33-3
EINECS: 205-358-3
Merck Index: 12,03556
现在订购
产品技术规格
For Laboratory,Research,or Manufacturing Use
Molarity 0.095 - 0.105