碘 0.01 N (N/100), StandARd® 滴定液, Macron Fine Chemicals™
供应商: Avantor Performance Materials
Management of Change (MOC) category = N
MACRH175-05EA 1227 CNY
MACRH175-05 MACRH175-07
碘 0.01 N (N/100), StandARd® 滴定液, Macron Fine Chemicals™
分子式: I₂
MDL编号: MFCD00011355
CAS号: 7553-56-2
现在订购
产品技术规格
For Laboratory,Research,or Manufacturing Use
Normality 0.0095 - 0.0105
Appearance (transparent, reddish-brown solution)