碘 0.10 N (N/10) in aqueous solution, StandARd® volumetric solution, Macron Fine Chemicals™
供应商: Avantor Performance Materials
Danger
MACR2962-07EA 2661 CNY
MACR2962-07 MACR2962-60
碘 0.10 N (N/10) in aqueous solution, StandARd® volumetric solution, Macron Fine Chemicals™
分子式: I₂
存储条件: Ambient
MDL编号: MFCD00011355
CAS号: 7553-56-2
EINECS: 231-442-4
ADR: 8,II
现在订购
产品技术规格
For Laboratory,Research,or Manufacturing Use 0.0995 - 0.1005
Normality 0.0995 - 0.1005
Appearance (dark brown to red-brown solution)