VWR®, Precision thermometers, liquid-in-glass
供应商: VWR BRAND PRIVATE LABEL USA
89095-822_U 89095-820
VWRU89095-822_UEA 0 CNY
VWRU89095-822_U VWRU89095-820
VWR®, Precision thermometers, liquid-in-glass
温度计 Analogue Thermometers
Precision thermometers are made with permanently fused markings and fractional degrees to provide precision and ensure accuracy.
  • Mercury-free, contain blue organic liquid against lead-free white back glass
  • Tested and inspected by our USA quality team
Ideal for use in chemical laboratories, universities, scientific institutions, food and beverage processing, petrochemical laboratories, pharmaceutical/medical industry, manufacturing, dairies, breweries and distilleries.
Certifications: Tested and calibrated in an exclusive triple accredited/registered ISO/IEC 17025:2005, A2LA accredited laboratory against equipment whose calibration is traceable to NIST, individually serialised.