Bags, Amber
供应商: Elkay Plastics
ELKAFAM30406EA 0 CNY
ELKAFAM30808 ELKAFAM30406 ELKAFAM31212 ELKA20FAM-0610
Bags, Amber
贮藏袋
Amber bags prevent their contents from being degraded or damaged by UV light waves.
  • Open-ended bags can be sealed with a heat sealer or taped shut
  • Seal top bags can be accessed multiple times
They are a choice for packaging and transporting medications and other UV light-sensitive products.