VWR® Performance, Refrigerators, for storage of flammable materials
供应商: VWR BRAND PRIVATE LABEL USA
10819-732 10819-734
VWRU10819-732EA 167419 CNY
VWRU10819-732 VWRU10819-734
VWR® Performance, Refrigerators, for storage of flammable materials
冰箱
Designed for the refrigerated storage of flammable or volatile materials in general laboratory work areas.
  • Data acquisition
  • Data logging 24/7
  • Digital temp chart (24-hour)
  • Min/max temp history
  • USB port for data transfer
No electrical components are located in the interior of the unit, and the electrical components of the compressor are sealed in a vapour-proof enclosure for additional safety. Units feature a digital microprocessor temperature controller that allows the precise temperature management necessary for critical samples and supplies.
Output: 4 to 20 mA
Certifications: Meet NFPA and OSHA guidelines 45 and 70. UL/C-UL listed.