VWR®Standard, Refrigerators, for storage of flammable material
供应商: VWR BRAND PRIVATE LABEL USA
10819-764 10819-766
VWRU10819-764EA 104197 CNY
VWRU10819-764 VWRU10819-766
VWR®Standard, Refrigerators, for storage of flammable material
冰箱
These Standard series refrigerators are designed for the storage of flammable or volatile materials in general laboratory work areas.
  • Adjustable operating temperature range: 1 to 10 °C
  • Electrical components are housed externally
No electrical components are located in the interior of the unit, and the electrical components of the compressor are sealed in a vapour-proof enclosure for additional safety. Standard features package includes keyed door locks, probe access port.
Units are supplied with a two years parts and labour warranty.
Certifications: Meets NFPA and OSHA guidelines, UL and C-UL listed.