VWR®, Serum Vials
供应商: VWR BRAND PRIVATE LABEL USA
89207-578 89207-576
VWRU89207-578EA 32605 CNY
VWRU89207-578 VWRU89207-576
VWR®, Serum Vials
小瓶
VWR® polycarbonate resin is an amorphous engineering thermoplastic, characterized by outstanding mechanical, optical, electrical, and thermal properties.
  • Highly durable
  • Resistant to scratches
  • Oustanding mechanical, optical, electrical, and thermal properties
  • Ideal for leak-proof shipment of reagents and buffer solutions
The Lexan® resins we employ are environmentally conforming to flame retardancy, scratch resistance, toughness, heat resistance, weatherability, biocompatibility, optical quality, and compliance with stringent FDA and USP requirements