VWR® Performance series, flammable material storage freezers
供应商: VWR BRAND PRIVATE LABEL USA
10819-692 10819-694
VWRU10819-692EA 202944 CNY
VWRU10819-692 VWRU10819-694
VWR® Performance series, flammable material storage freezers
冷柜 实验室通用冰箱
The VWR® performance series of flammable material freezers are designed for refrigerated storage of flammable or volatile materials in general laboratory work areas.
  • Data acquisition
  • 24/7 data logging
  • 24-hour digital temp chart
Units are equipped with USB port for data transfer, 4 - 20 mA output, min/max temp history, digital microprocessor temperature controller allowing for the precise temperature management necessary for critical samples and supplies. No electrical components are located in the interior of the unit, and the electrical components of the compressor are sealed in a vapour-proof enclosure for additional safety.
Certifications: Units meet NFPA and OSHA guidelines 45 and 70, and are UL and C-UL listed.