VWR®, Cellulose Sponges, Fine-Pore
供应商: VWR BRAND PRIVATE LABEL USA
58542-360 58542-246
VWRU58542-360EA 0 CNY
VWRU58542-360 VWRU58542-246
VWR®, Cellulose Sponges, Fine-Pore
擦拭巾 海绵
These fine pore natural colour cellulose sponges hold approximately 25 times their own weight in water.
  • Durable
  • Convenient for quick laboratory wipe-up
Resist tearing and abrasion. Absorb liquid more evenly than large pore sponges.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904