VWR®, Protective gowns, SPP
供应商: VWR BRAND PRIVATE LABEL USA
414004-467 414004-468 89178-402_U
VWRU414004-467EA 992 CNY
VWRU414004-467 VWRU414004-468 VWRU89178-402_U
VWR®, Protective gowns, SPP
Disposable Garments
Economical gowns are manufactured from a strong, lightweight, breathable and non-linting spunbonded polypropylene (SPP) fabric.
  • Provide first line barrier
  • Elastic cuffs and ties at the neck and waist ensure secure closure with
  • Serged seams
  • No pockets
Gowns protect against dirt, grime and certain dry particulates in nonhazardous environments.
Certifications: Tested and manufactured in an ISO certified facility under stringent process controls. Lot-controlled and validated through independent laboratory testing.