VWR®, Shoe covers for basic protection, SPP
供应商: VWR BRAND PRIVATE LABEL USA
97041-236 414004-653 97041-234
VWRU97041-236EA 672 CNY
VWRU97041-236 VWRU414004-653 VWRU97041-234
VWR®, Shoe covers for basic protection, SPP
Disposable Garments
These economical shoe covers are made from low linting spunbonded polypropylene (SPP). The fabric is strong, lightweight and breathable.
  • Provide first-line barrier
  • Elastic bands on top and bottom
  • Available with rubber-coated soles for secure footing on wet or dry surfaces
Shoe covers protect against dirt, grime, and certain dry particulates in non-hazardous environments. Appropriate for industrial, pharmaceutical, food processing, construction and certain environmental cleanup applications.
Certifications: Tested and manufactured in an ISO certified facility under stringent process controls. Lot-controlled and validated through independent laboratory testing.