VWR®, Pinch Clamps
供应商: VWR BRAND PRIVATE LABEL USA
89259-872 89259-878 89259-884
VWRU89259-872EA 327 CNY
VWRU89259-872 VWRU89259-878 VWRU89259-884
VWR®, Pinch Clamps
夹具 通用夹具
Pinch clamps for glass ball and socket or O-ring joints add convenience and speed when mounting and dismantling a glass apparatus.
  • Corrosion-resistant
  • Stainless steel construction
  • Fast and convenient mounting and dismantling of glass apparatuses
Clamps have strong spring-closed forked jaws and a screw locking device.