VWR®, Tube Rockers Collection
供应商: VWR BRAND PRIVATE LABEL USA
10159-754 10159-752
VWRU10159-754EA 0 CNY
VWRU10159-754 VWRU10159-752
VWR®, Tube Rockers Collection
振荡器 旋转/ 摇摆式振荡器
The VWR® tube rocker collection features mini as well as an XL size tube rockers and variable speed models.
  • Low profile and small footprint
  • Tube rockers provide smooth, gentle mixing
  • Available with a variety of speed settings
The mini tube rockers holds 8 tubes, 120 mm long while the XL model holds up to 18 tubes, also 120 mm long. Each model is equipped with a fixed tilt angle of 35° and mixes your samples simultaneously via a 110 to 240 V power pack. The variable speed models are adjustable in 1 rpm increments from 5 to 25 rpm while the fixed speed model is fixed at 18 rpm.
Certifications: UL/CSA approved.
Delivery information: Supplied with a 12 V AC adapter.