VWR® SuperSlips™, Micro Cover Glasses, Rectangular
供应商: VWR BRAND PRIVATE LABEL USA
48404-455 48404-453
VWRU48404-455EA 0 CNY
VWRU48404-455 VWRU48404-453
VWR® SuperSlips™, Micro Cover Glasses, Rectangular
盖玻片
These cover glasses are corrosion-resistant and uniformly thin and flat.
  • Suitable for humid environments
  • Resistant to corrosion
  • Processing techniques result in superior cleanliness
SuperSlip coverslips combine specialised production methods with moisture-resistant packaging to prevent sticking.