VWR®, Overalls with boots, SMS
供应商: VWR BRAND PRIVATE LABEL USA
414004-687 414004-688 414004-689 414004-690 414004-691 414004-692 414004-693 414004-694
VWRU414004-687EA 2246 CNY
VWRU414004-687 VWRU414004-688 VWRU414004-689 VWRU414004-690 VWRU414004-691 VWRU414004-692 VWRU414004-693 VWRU414004-694
VWR®, Overalls with boots, SMS
Disposable Garments
These overalls with integrated shoe covers are manufactured from a unique multilayered spunbonded-meltblown-spunbonded (SMS) fabric that offers high tensile strength while remaining soft, comfortable and breathable. The material is low-linting and resistant to tears and punctures.
  • Light fluid and particulate barrier
  • Excellent breathability
  • Inset sleeves, front zipper, tapered collar
  • Elastic cuffs, ankles, and back
  • No pockets
  • Serged seams
These static-dissipative garments act as a fluid and particulate barrier and provide light chemical splash resistance. Ideal for use in high-class protection areas of medical, industrial, laboratory, electronic, and pharmaceutical environments.
Certifications: Tested and manufactured in an ISO certified facility under stringent process controls. Lot-controlled and validated through independent laboratory testing.