1,3,5-三羟基苯, J.T.Baker®

供应商: Avantor
Warning

Synonyms: 间苯三酚

BAKRU024-05EA 2981 CNY
BAKRU024-05
1,3,5-三羟基苯, J.T.Baker®
1,3,5-三羟基苯
分子式: C₆H₆O₃·2H₂O
分子量: 162.14 g/mol
溶点: 213…220 °C
存储条件: Ambient
MDL编号: MFCD00149090
CAS号: 6099-90-7
EINECS: 203-611-2
Merck Index: 13,07413

现在订购

产品技术规格

For Laboratory,Research,or Manufacturing Use
Melting Point 213 - 220 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR