氨水 分析级 5 N, BAKER ANALYZED® Volumetric, J.T.Baker®
供应商: Avantor
Synonyms: Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
M.O.C. Category N
BAKR5604-02EA 1197 CNY
BAKR5604-02 BAKR5604-07
氨水 分析级 5 N, BAKER ANALYZED® Volumetric, J.T.Baker®
氨水 分析级
分子式: NH₃
分子量: 35.05 g/mol
密度: 0.90 g/cm³
存储条件: Ambient
MDL编号: MFCD00066650
CAS号: 1336-21-6
EINECS: 215-647-6
UN号: 2672
ADR: 8 III
现在订购
产品技术规格
For Laboratory,Research,or Manufacturing Use
Normality 4.95 - 5.05
Standardization at 25°C traceable to NIST Standard Reference Material.
Normality should be confirmed at the time of use because Ammonium
Hydroxide may vaporize thus reducing its normality.