氢氧化钠 ≥98% (by acidimetry), pellets, BAKER ANALYZED® ACS, J.T.Baker®
供应商: Avantor
|
Danger
|
Synonyms:
苛性钠
Management of Change (MOC) category = N
BAKR3115-01EA
323
CNY
BAKR3115-01
BAKR3115-05
氢氧化钠 ≥98% (by acidimetry), pellets, BAKER ANALYZED® ACS, J.T.Baker®
氢氧化钠
分子式:
NaOH 分子量: 40 g/mol 沸点: 1390 溶点: 323 |
MDL编号:
MFCD00003548 CAS号: 1310-73-2 UN号: 1823 ADR: 8 II Merck Index: 14,08627 |
产品技术规格
For Laboratory,Research,or Manufacturing Use | |
Assay (NaOH) (by acidimetry) | ≥98 % |
Calcium (Ca) | ≤0.005 % |
Chloride (Cl) | ≤0.005 % |
Copper (Cu) | ≤0.001 % |
Heavy Metals (as Ag) | ≤0.001 % |
Insoluble Matter | ≤0.003 % |
Iron (Fe) | ≤0.0003 % |
ACS - Magnesium (Mg) | ≤0.002 % |
Mercury (Hg) | ≤0.1 ppm |
Nickel (Ni) | ≤0.0005 % |
Nitrogen Compounds (as N) | ≤0.0003 % |
Phosphate (PO₄) | ≤0.0002 % |
Potassium (K) | ≤0.02 % |
Sodium Carbonate (Na₂CO₃) | ≤0.4 % |
ACS - Sulfate (SO₄) | ≤0.003 % |
Meets Reagent Specifications for testing USP/NF monographs | |
Appearance (white hygroscopic pellets) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...