Accutase®
供应商: Merck
A6964-100ML A6964-500ML
SIALA6964-100MLEA 0 CNY
SIALA6964-100ML SIALA6964-500ML
Accutase®
Accutase is supplied as a sterile filtered solution suitable for cell culture.
  • Ready to use
  • Does not contain mammalian or bacterial-derived products
Used in detaching cells for analysis of cell surface markers, virus growth assay, and flow cytometry as well as bioreactor scale-up. Efficient in detaching primary fibroblasts, endothelial cells, neurons, tumor cell lines, and insect cells.