PCR tube strips, 8-well, low profile
供应商: BIO RAD
TLS-0801 TLS-0851
BRADTLS-0801EA 0 CNY
BRADTLS-0801 BRADTLS-0851
PCR tube strips, 8-well, low profile
管子 聚合酶链反应管
Thin wall PP 8-tube strips for PCR and low-volume reactions.
  • Low overall height (15,48 mm) reduces condensation
  • Ideal for fast PCR, low volume reactions, and real-time PCR detection systems
  • Free from DNase, RNase, and human DNA