三氯异氰尿酸 99%

供应商: Thermo Fisher Scientific
Danger

Synonyms: TCICA, 三氯氰尿酸, 三氯三嗪三酮, 强氯精

421540010 421542500
ACRO421540010EA 0 CNY
ACRO421540010 ACRO421542500
三氯异氰尿酸 99%
三氯异氰尿酸
分子式: C₃Cl₃N₃O₃
分子量: 232.41 g/mol
沸点: 413
溶点: 245…251 °C
存储条件: Ambient
MDL编号: MFCD00006553
CAS号: 87-90-1
EINECS: 201-782-8
UN号: 2468
ADR: 5.1 II
Merck Index: 12,09188

现在订购

产品技术规格

Appearance White to off-white Powder or granules
Infrared spectrum Conforms
Titration Iodometric ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR