硫代氨基脲 98+%

供应商: Thermo Fisher Scientific
Danger

Synonyms: 肼基碳硫酰胺

138901000 138905000
ACRO138901000EA 0 CNY
ACRO138901000 ACRO138905000
硫代氨基脲 98+%
硫代氨基脲
分子式: CH₅N₃S
分子量: 91.14 g/mol
溶点: 175…180 °C
存储条件: Ambient
MDL编号: MFCD00007620
CAS号: 79-19-6
EINECS: 201-184-7
UN号: 2811
ADR: 6.1 II
Merck Index: 12,09499

现在订购

产品技术规格

Appearance White to yellow to beige Powder or crystals
Infrared spectrum Conforms
Melting point 175 to 180 °C
Titration with HClO4 ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR