2-巯基嘧啶 98%
供应商: Thermo Fisher Scientific
Synonyms:
2-嘧啶硫醇
156740100
156740500
ACRO156740100EA
0
CNY
ACRO156740100
ACRO156740500
2-巯基嘧啶 98%
2-巯基嘧啶
分子式:
C₄H₄N₂S 溶点: 230 °C 存储条件: Ambient |
MDL编号:
MFCD00006073 CAS号: 1450-85-7 EINECS: 215-917-3 |
产品技术规格
Appearance | Yellow Powder |
Infrared spectrum | Conforms |
Titration with NaOH | ≥97.5 % |
Phosphate (PO4) | ≤5 ppm |
Ammonium (NH4) | ≤5 ppm |
Calcium (Ca) | ≤0.002 % |
Iron (Fe) | ≤5 ppm |
Magnesium (Mg) | ≤0.001 % |
Sodium (Na) | ≤0.03 % |
Heavy metals (as Pb) | ≤5 ppm |
Sulfur compounds (as SO4) | ≤0.003 % |
Insoluble matter | ≤0.01 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...