2-(2-吡啶基)乙胺 95%
供应商: Thermo Fisher Scientific
|
Danger
|
Synonyms:
2-(2-氨乙基)吡啶
103690100
103690010
103690500
ACRO103690100EA
0
CNY
ACRO103690100
ACRO103690010
ACRO103690500
2-(2-吡啶基)乙胺 95%
2-(2-吡啶基)乙胺
分子式:
C₇H₁₀N₂ 分子量: 122.17 g/mol 沸点: 105…106 °C (10 mmHg) 密度: 1.021 g/cm³ 闪点: 100 °C 存储条件: Ambient |
MDL编号:
MFCD00006367 CAS号: 2706-56-1 EINECS: 220-295-1 UN号: 2735 |
产品技术规格
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5340 to 1.5380 (20°C, 589 nm) |
GC | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...