2-氨基-5-硝基吡啶 99%
供应商: Thermo Fisher Scientific
|
Warning
|
Synonyms:
5-Nitropyridin-2-ylamine, 5-Nitropyridin-2-amine, 5-Nitro-2-pyridylamine
104230250
104231000
104230050
ACRO104230250EA
0
CNY
ACRO104230250
ACRO104231000
ACRO104230050
2-氨基-5-硝基吡啶 99%
2-氨基-5-硝基吡啶
分子式:
C₅H₅N₃O₂ 分子量: 139.11 g/mol 溶点: 186…190 °C 存储条件: Ambient |
MDL编号:
MFCD00006325 CAS号: 4214-76-0 EINECS: 224-145-6 |
产品技术规格
Appearance | Yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 186 to 190 °C |
HPLC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...