2-羟基吡啶-3-羧酸 98%

供应商: Thermo Fisher Scientific
Warning

Synonyms: 2-羟基吡啶-3-羧酸, 2-羟基烟酸

247220250 247221000
ACRO247220250EA 0 CNY
ACRO247220250 ACRO247221000
2-羟基吡啶-3-羧酸 98%
2-羟基吡啶-3-羧酸
分子式: C₆H₅NO₃
分子量: 139.11 g/mol
溶点: 259…261 °C
MDL编号: MFCD00010100
CAS号: 609-71-2
EINECS: 210-198-2

现在订购

产品技术规格

Appearance White to cream to light yellow Powder
Infrared spectrum Conforms
Melting point 256 to 262 °C
Titration with NaOH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR