2-氯-3-吡啶甲腈 ≥97.0% (by GC)
供应商: TCI
|
Danger
|
Synonyms:
2-氯-3-氰基吡啶
C1369-25G
C1369-500G
TCIAC1369-25GEA
0
CNY
TCIAC1369-25G
TCIAC1369-500G
2-氯-3-吡啶甲腈 ≥97.0% (by GC)
2-氯-3-吡啶甲腈
分子式:
C₆H₃ClN₂ 分子量: 138.56 g/mol 沸点: 112 |
MDL编号:
MFCD00014628 CAS号: 6602-54-6 EINECS: 229-550-1 UN号: 3439 ADR: 6.1 III |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 105.0 to 109.0 deg-C |
Solubility in hot Methanol | very faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...