5-溴-2-氟吡啶 99%
供应商: Thermo Fisher Scientific
|
Danger
|
378460050
378460250
ACRO378460050EA
0
CNY
ACRO378460050
ACRO378460250
5-溴-2-氟吡啶 99%
5-溴-2-氟吡啶
分子式:
C₅H₃BrFN 分子量: 175.99 g/mol 沸点: 63 °C (15 mmHg) 闪点: 73 °C |
MDL编号:
MFCD01863742 CAS号: 766-11-0 UN号: 2810 ADR: 6.1 III |
产品技术规格
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5310 to 1.5330 (20°C, 589 nm) |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...