3-Methyladenine 90+%

供应商: Thermo Fisher Scientific
Warning

Synonyms: 3-Methyl-3H-purin-6-amine, 3-Methyl-3H-adenine

379791000 379795000
ACRO379791000EA 0 CNY
ACRO379791000 ACRO379795000
3-Methyladenine 90+%
3-Methyladenine
分子式: C₆H₇N₅
分子量: 149.16 g/mol
溶点: 300 °C
存储条件: Refrigerator
MDL编号: MFCD00010531
CAS号: 5142-23-4
EINECS: 225-908-6

现在订购

产品技术规格

Appearance White Powder
Infrared spectrum Conforms
HPLC ≥90.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR