次黄嘌呤 99.5%

供应商: Thermo Fisher Scientific
Warning

Synonyms: 6-羟基嘌呤

122010250 122010050
ACRO122010250EA 0 CNY
ACRO122010250 ACRO122010050
次黄嘌呤 99.5%
次黄嘌呤
分子式: C₅H₄N₄O
分子量: 136.11 g/mol
溶点: >250 °C (dec.)
存储条件: Ambient
MDL编号: MFCD00005725
CAS号: 68-94-0
EINECS: 200-697-3
Merck Index: 13,04895

现在订购

产品技术规格

Appearance White to light yellow to light beige Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤0.5 %
Sulfated ash ≤0.2 %
HPLC ≥99.4 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR