氰化锌 98%, pure
供应商: Thermo Fisher Scientific
|
Danger
|
Synonyms:
Zinc (II) cyanide
368070050
368072500
368070010
ACRO368070050EA
0
CNY
ACRO368070050
ACRO368072500
ACRO368070010
氰化锌 98%, pure
氰化锌
分子式:
C₂N₂Zn 分子量: 117.43 g/mol 溶点: 800 密度: 1.852 存储条件: Ambient |
MDL编号:
MFCD00011292 CAS号: 557-21-1 EINECS: 209-162-9 UN号: 1713 ADR: 6.1 I Merck Index: 12,10264 |
产品技术规格
Appearance | White to off-white Powder |
Titration Complexometric | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...