5-甲氧基吲哚 99%

供应商: Thermo Fisher Scientific
Warning

Synonyms: 5-Methoxy-1H-indole, Indol-5-yl methyl ether

125870050 125870250
ACRO125870050EA 0 CNY
ACRO125870050 ACRO125870250
5-甲氧基吲哚 99%
5-甲氧基吲哚
分子式: C₉H₉NO
分子量: 147.18 g/mol
沸点: 176…178 °C (17 mmHg)
溶点: 52…56 °C
存储条件: Refrigerator
MDL编号: MFCD00005674
CAS号: 1006-94-6
EINECS: 213-745-3

现在订购

产品技术规格

Appearance White to brown Powder
Infrared spectrum Conforms
Melting point 52 to 56 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR