2-甲基咪唑 99%
供应商: Thermo Fisher Scientific
|
Danger
|
Synonyms:
2-甲基-1,3-氮杂茂
127000010
127001000
127005000
ACRO127000010EA
0
CNY
ACRO127000010
ACRO127001000
ACRO127005000
2-甲基咪唑 99%
2-甲基咪唑
分子式:
C₄H₆N₂ 分子量: 82.11 g/mol 沸点: 267 溶点: 142…145 °C 闪点: 155 存储条件: Ambient |
MDL编号:
MFCD00005190 CAS号: 693-98-1 EINECS: 211-765-7 UN号: 3263 ADR: 8 III |
产品技术规格
Appearance | White to light yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 142 to 145 °C |
Water | ≤0.5 % (K.F.) |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...