4-硝基咪唑 98%
供应商: Thermo Fisher Scientific
|
Warning
|
Synonyms:
4-Nitro-1H-imidazole, 5-Nitroimidazole, 5-Nitro-1H-imidazole
161600250
161601000
ACRO161600250EA
0
CNY
ACRO161600250
ACRO161601000
4-硝基咪唑 98%
4-硝基咪唑
分子式:
C₃H₃N₃O₂ 分子量: 113.08 g/mol 溶点: 303 °C (dec.) 闪点: >200 °C 存储条件: Ambient |
MDL编号:
MFCD00005196 CAS号: 3034-38-6 EINECS: 221-224-7 |
产品技术规格
Appearance | White to light yellow Powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Titration with NaOH | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...