二甲基亚砜 99.9+% ACS

供应商: Thermo Fisher Scientific

Synonyms: 甲基亚砜, DMSO

414885000 414880010
ACRO414885000EA 0 CNY
ACRO414885000 ACRO414880010
二甲基亚砜 99.9+% ACS
二甲基亚砜
分子式: (CH₃)₂SO
沸点: 189 °C (760 mmHg)
密度: 1,1 g/cm³
闪点: 87 °C
存储条件: Ambient
MDL编号: MFCD00002089
CAS号: 67-68-5
EINECS: 200-664-3
Merck Index: 13,03285
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ec1573e80c11ddf9254283ef76abf5fb6539c7e&vid=157e2a24c93041cbbb69833d226195e333b07665&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Clear colorless Liquid
Water ≤0.1 % (Coulometric)
GC ≥99.9 %
Residue after evaporation ≤0.01 %
Titrable acid ≤0.001 meq/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR