氯化硫酰 98.5%
供应商: Thermo Fisher Scientific
|
Danger
|
Synonyms:
Sulphuryl dichloride
169450010
169450025
169450250
ACRO169450010EA
0
CNY
ACRO169450010
ACRO169450025
ACRO169450250
氯化硫酰 98.5%
氯化硫酰
分子式:
Cl₂O₂S 沸点: 69,1 °C (760 mmHg) 密度: 1,667 g/cm³ 存储条件: Ambient |
MDL编号:
MFCD00011451 CAS号: 7791-25-5 EINECS: 232-245-6 UN号: 1834 ADR: 6.1,I Merck Index: 12,09153 |
产品技术规格
Appearance | Clear colorless to yellow Liquid |
Titration Argentometric | ≥98.0 % (ex Cl) |
Free chlorine | ≤0.1 % |
Sulfur dioxide (SO2) | ≤0.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...