四氟硼酸铵 99%, pure

供应商: Thermo Fisher Scientific
Danger

Synonyms: 氟硼酸铵

310961000 310965000
ACRO310961000EA 0 CNY
ACRO310961000 ACRO310965000
四氟硼酸铵 99%, pure
四氟硼酸铵
分子式: H₄BF₄N
溶点: 230 °C (subl.)
密度: 1,87 g/cm³ (20 °C)
存储条件: Ambient
MDL编号: MFCD06245785
CAS号: 13826-83-0
EINECS: 237-531-4
UN号: 3260
ADR: 8,III

现在订购

产品技术规格

Appearance White Crystalline powder or crystals
Titration with NaOH ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR