四丙基溴化铵 98%
供应商: Thermo Fisher Scientific
|
Warning
|
Synonyms:
TPABr
209370250
209371000
209375000
ACRO209370250EA
0
CNY
ACRO209370250
ACRO209371000
ACRO209375000
四丙基溴化铵 98%
四丙基溴化铵
分子式:
C₁₂H₂₈BrN 分子量: 266.27 g/mol 溶点: 266…272 °C 存储条件: Ambient |
MDL编号:
MFCD00011840 CAS号: 1941-30-6 EINECS: 217-727-6 |
产品技术规格
Appearance | White to light yellow Powder or crystals |
Infrared spectrum | Conforms |
Titration Argentometric | 97.5 to 102.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...