四氟硼酸四乙胺 99%
供应商: Thermo Fisher Scientific
|
Warning
|
Synonyms:
Tetrylammonium tetrafluoroborate
221610250
221611000
ACRO221610250EA
0
CNY
ACRO221610250
ACRO221611000
四氟硼酸四乙胺 99%
四氟硼酸四乙胺
分子式:
C₈H₂₀BF₄N 溶点: >300 °C 存储条件: Ambient |
MDL编号:
MFCD00011827 CAS号: 429-06-1 EINECS: 207-055-1 ADR: 6.1,III |
产品技术规格
Appearance | White Crystalline powder or crystals |
Infrared spectrum | Conforms |
Solubility | (4 % in acetonitrile) Clear to almost clear colorless |
Elemental Analysis | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...