乙炔基三甲基硅烷 98%

供应商: Thermo Fisher Scientific
Danger

Synonyms: 三甲基硅乙炔

203570050 203570250
ACRO203570050EA 0 CNY
ACRO203570050 ACRO203570250
乙炔基三甲基硅烷 98%
乙炔基三甲基硅烷
分子式: C₅H₁₀Si
沸点: 53 °C (1013 hPa)
密度: 0,69 g/cm³
闪点: –34 °C
存储条件: Refrigerator
MDL编号: MFCD00008569
CAS号: 1066-54-2
EINECS: 213-919-9
UN号: 1993
ADR: 3,II

现在订购

产品技术规格

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3873 to 1.3903 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR