硼酸三丁酯 98%
供应商: Thermo Fisher Scientific
|
Warning
|
Synonyms:
硼酸三正丁酯, 三丁氧基甲硼烷
177191000
177195000
ACRO177191000EA
0
CNY
ACRO177191000
ACRO177195000
硼酸三丁酯 98%
硼酸三丁酯
分子式:
C₁₂H₂₇BO₃ 分子量: 230.15 g/mol 沸点: 230…235 °C (760 mmHg) 溶点: –70 °C 密度: 0.853 g/cm³ 闪点: 93 °C 存储条件: Ambient |
MDL编号:
MFCD00009434 CAS号: 688-74-4 EINECS: 211-706-5 |
产品技术规格
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4080 to 1.4100 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...