碘化铟(III) 99.998% (trace metals basis)
供应商: Thermo Fisher Scientific
|
Danger
|
316010010
316010050
ACRO316010010EA
0
CNY
ACRO316010010
ACRO316010050
碘化铟(III) 99.998% (trace metals basis)
碘化铟(III)
分子式:
I₃In 分子量: 495.53 g/mol 沸点: 500 溶点: 210 °C 存储条件: Ambient |
MDL编号:
MFCD00016151 CAS号: 13510-35-5 EINECS: 236-839-6 |
产品技术规格
Appearance | Yellow to light brown Powder or crystals |
Titration Complexometric | 22.7 to 23.5 % (In) |
Total trace metal impurities | ≤0.002 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...