三溴化硼 99+%

供应商: Thermo Fisher Scientific
Danger

365210250 365211000
ACRO365210250EA 0 CNY
ACRO365210250 ACRO365211000
三溴化硼 99+%
三溴化硼
分子式: BBr₃
沸点: 91,2 °C (760 mmHg)
溶点: –46 °C
密度: 2,65 g/cm³
存储条件: Ambient
MDL编号: MFCD00011312
CAS号: 10294-33-4
EINECS: 233-657-9
UN号: 2692
ADR: 8,I
Merck Index: 13,01337

现在订购

产品技术规格

Appearance Clear amber Liquid
Titration ≥99.0 %
Sulfur (S) ≤100 ppm
Heavy metals (as Pb) ≤100 ppm
Impurity ≤0.2 % Free bromine (Br2)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR