六氰基铁(III)酸钾 99+% ACS

供应商: Thermo Fisher Scientific

Synonyms: 赤血盐, 铁氰化钾

424120050 424125000
ACRO424120050EA 0 CNY
ACRO424120050 ACRO424125000
六氰基铁(III)酸钾 99+% ACS
六氰基铁(III)酸钾
分子式: K₃Fe(CN)₆
分子量: 329.25 g/mol
存储条件: Ambient
MDL编号: MFCD00011392
CAS号: 13746-66-2
EINECS: 237-323-3
Merck Index: 13,07712

现在订购

产品技术规格

Appearance Orange to red Crystals and/or chunks
Titration Iodometric ≥99.0 %
Chloride (Cl) ≤0.01 %
Sulfate (SO4) ≤0.01 %
Ferro compounds ≤0.05 % as ferrocyanide ion ¢Fe(CN)6!4-
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR