亚氯酸钠 80% unstabilised, 纯净

供应商: Thermo Fisher Scientific
Danger

223230050 223235000
ACRO223230050EA 0 CNY
ACRO223230050 ACRO223235000
亚氯酸钠 80% unstabilised, 纯净
亚氯酸钠
分子式: ClNaO₂
沸点: 112 °C (1013 hPa)
溶点: 180…200 °C (dec.)
密度: 1,28 g/cm³ (20 °C)
存储条件: Ambient
MDL编号: MFCD00003478
CAS号: 7758-19-2
EINECS: 231-836-6
UN号: 1496
ADR: 5.1,II
Merck Index: 13,08672

现在订购

产品技术规格

Appearance White Powder
Titration Iodometric ≥75 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR