氯化钠, Sigma-Aldrich®

供应商: Merck


M-Clarity™ quality level = MQ200

S7653-1KG S7653-5KG
SIALS7653-1KGEA 0 CNY
SIALS7653-1KG SIALS7653-5KG
氯化钠, Sigma-Aldrich®
氯化钠
分子式: NaCl
沸点: 1413 °C (1013 hPa)
溶点: 801 °C
密度: 2,16 g/cm³ (20 °C)
存储条件: Ambient
MDL编号: MFCD00003477
CAS号: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,08599

现在订购


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR